| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10461254 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10461252 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10461253 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10461255 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1437771_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10461251 |
- |
- |
| Mouse Genome 430 2.0 Array |
1437771_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1437771_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
68852_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
68852_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1900002158.mm.1 |
NM_001013019.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002158.mm.1 |
NM_001013019.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002158.mm.1 |
TC1900000161.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002152.mm.1 |
NM_001013019.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002152.mm.1 |
NM_001013019.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002152.mm.1 |
TC1900000161.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002153.mm.1 |
NM_001013019.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002153.mm.1 |
NM_001013019.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002153.mm.1 |
TC1900000161.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002156.mm.1 |
NM_001013019.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002156.mm.1 |
NM_001013019.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002156.mm.1 |
TC1900000161.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002157.mm.1 |
NM_001013019.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002157.mm.1 |
NM_001013019.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002157.mm.1 |
TC1900000161.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002154.mm.1 |
NM_001013019.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002154.mm.1 |
NM_001013019.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002154.mm.1 |
TC1900000161.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002155.mm.1 |
NM_001013019.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002155.mm.1 |
NM_001013019.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002155.mm.1 |
TC1900000161.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1437771_at |
- |
- |
| Mouse Expression Set 430 |
1437771_PM_at |
- |
- |
| Murine Genome U74v2 Set |
166520_r_at |
- |
- |
| Murine Genome U74v2 Set |
166520_PM_r_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1437771_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1437771_PM_at |
- |
- |