Assay Details
| Gene Symbol: Yif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026553.4 | NP_080829.1 | 2 - 3 | 427 | 66 | |
| GenBank mRNA | AK011071.1 | - | 2 - 3 | 421 | 66 | |
| AK077329.1 | - | 2 - 3 | 413 | 66 | ||
| AK160409.1 | - | 2 - 3 | 380 | 66 | ||
| BC011117.1 | - | 2 - 3 | 369 | 4190535 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yif1a
Genomic Map