Assay Details
| Gene Symbol: Ralbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198949.1 | NP_001185878.1 | 7 - 8 | 1757 | 159 | |
| NM_009067.5 | NP_033093.2 | 7 - 8 | 2081 | 159 | ||
| GenBank mRNA | AK145387.1 | - | 7 - 8 | 1621 | 159 | |
| AK159404.1 | - | 7 - 8 | 1634 | 159 | ||
| BC054329.1 | - | 7 - 8 | 1672 | 159 | ||
| BC067073.1 | - | 6 - 7 | 1516 | 30611880 | 159 | |
| BC075732.1 | - | 7 - 8 | 1706 | 6507517 | 159 | |
| BC076636.1 | - | 7 - 8 | 1673 | 30658617 | 159 | |
| BC138546.1 | - | 6 - 7 | 1522 | 8861566 | 159 | |
| BC145320.1 | - | 5 - 6 | 1360 | 9053875 | 159 | |
| BC145815.1 | - | 6 - 7 | 1522 | 40131118 | 159 | |
| X80937.1 | - | 7 - 8 | 1658 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ralbp1
Genomic Map