Assay Details
| Gene Symbol: Tex264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081654.2 | NP_001075123.1 | 4 - 5 | 707 | 106 | |
| NM_001286498.1 | NP_001273427.1 | 4 - 5 | 907 | 106 | ||
| NM_011573.3 | NP_035703.2 | 4 - 5 | 1162 | 106 | ||
| NR_104459.1 | 3 - 4 | 393 | 106 | |||
| GenBank mRNA | AK009326.1 | - | 4 - 5 | 1013 | 106 | |
| BC002248.1 | - | 4 - 5 | 671 | 3493141 | 106 | |
| BC006608.1 | - | 4 - 5 | 645 | 3157453 | 106 | |
| DV656490.1 | - | 3 - 4 | 360 | 106 | ||
| DV661702.1 | - | 2 - 3 | 482 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tex264
Genomic Map