Assay Details
| Gene Symbol: Col6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172927.3 | NP_766515.2 | 1 - 2 | 221 | 61 | |
| GenBank mRNA | AK054356.1 | - | 1 - 2 | 222 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK054356.1, NM_172927.3 transcripts.
Set Membership: |
|

Col6a6
Genomic Map