| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00017590.mm.1 |
NM_009197.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017590.mm.1 |
NM_009197.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017590.mm.1 |
TC0X00002783.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017592.mm.1 |
NM_009197.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017592.mm.1 |
NM_009197.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017592.mm.1 |
TC0X00002783.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017591.mm.1 |
NM_009197.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017591.mm.1 |
NM_009197.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017591.mm.1 |
TC0X00002783.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017588.mm.1 |
NM_009197.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017588.mm.1 |
NM_009197.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017588.mm.1 |
TC0X00002783.mm.1 |
MTA TC |