Assay Details
| Gene Symbol: Tmem263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013028.2 | NP_001013046.1 | 2 - 3 | 439 | 101 | |
| GenBank mRNA | AK005698.1 | - | 324 | 101 | ||
| AK142035.1 | - | 2 - 3 | 292 | 101 | ||
| BC086492.1 | - | 2 - 3 | 304 | 30470653 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005698.1, AK142035.1, BC086492.1, NM_001013028.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27306492 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27306489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27306493 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P545643 | BC086492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P545643 | NM_001013028.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P545643 | BC086492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P545643 | NM_001013028.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13914 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178419 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121545 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165529 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504775 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108887 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96192 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533780 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18183 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172175 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P545643 | BC086492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P545643 | NM_001013028.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P545643 | BC086492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P545643 | NM_001013028.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306494 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172175 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07133084 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07133067 | - | - |
| Whole Mouse Genome G4122A | A_52_P545643 | BC086492.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P545643 | NM_001013028.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P545643 | BC086492.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P545643 | NM_001013028.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100240 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07133067 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172175 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08303233 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08303239 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27306489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27306493 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27306492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08229677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08387129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08303241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08303237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08303233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306546 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10365421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365422 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431280_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433897_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10365420 | - | - |
| Mouse Genome 430 2.0 Array | 1433897_at | - | - |
| Mouse Genome 430 2.0 Array | 1433897_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431280_at | - | - |
| Mouse Genome 430 2.0 Array | 1431280_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103266_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103266_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000007768.mm.1 | NM_001013028.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007768.mm.1 | NM_001013028.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007768.mm.1 | TC1000001027.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000007767.mm.1 | NM_001013028.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007767.mm.1 | NM_001013028.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007767.mm.1 | TC1000001027.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000007766.mm.1 | NM_001013028.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007766.mm.1 | NM_001013028.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000007766.mm.1 | TC1000001027.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431280_at | - | - |
| Mouse Expression Set 430 | 1431280_PM_at | - | - |
| Mouse Expression Set 430 | 1433897_at | - | - |
| Mouse Expression Set 430 | 1433897_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431280_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431280_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431280_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431280_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433897_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433897_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105290152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105290152 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160449 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160449 | - | - |
| mouse-6 v1.1 (Illumina) | GI_61557490-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105290152 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105290152 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160449 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160449 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213317 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_61557490-S | - | - |

Tmem263
Genomic Map