Assay Details
| Gene Symbol: Crim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015800.3 | NP_056615.1 | 1 - 2 | 855 | 91 | |
| GenBank mRNA | AF168680.1 | - | 1 - 2 | 307 | 91 | |
| BC100348.1 | - | 1 - 2 | 640 | 30521148 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crim1
Genomic Map