Assay Details
| Gene Symbol: Crim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015800.3 | NP_056615.1 | 8 - 9 | 2022 | 57 | |
| GenBank mRNA | AF168680.1 | - | 8 - 9 | 1474 | 57 | |
| AK053197.1 | - | 6 - 7 | 965 | 57 | ||
| BC100348.1 | - | 8 - 9 | 1807 | 30521148 | 57 | |
More Information
Set Membership: |
|

Crim1
Genomic Map