Assay Details
| Gene Symbol: Mterf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028832.3 | NP_083108.1 | 1 - 2 | 62 | 101 | |
| GenBank mRNA | AK047778.1 | - | 1 - 2 | 38 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK047778.1, NM_028832.3 transcripts.
Set Membership: |
|

Mterf2
Genomic Map