Assay Details
| Gene Symbol: Mterf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028832.3 | NP_083108.1 | 2 - 3 | 180 | 93 | |
| GenBank mRNA | AK047778.1 | - | 2 - 3 | 156 | 93 | |
| BC046619.1 | - | 1 - 2 | 127 | 6494011 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK047778.1, BC046619.1, NM_028832.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202623 | NM_028832.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202623 | AA285696.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202623 | AK047778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202623 | BC046619.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202623 | NM_028832.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202623 | AA285696.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202623 | AK047778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202623 | BC046619.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156844 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 26325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25732 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659022 | AK039328.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157882 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145706 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202623 | NM_028832.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202623 | AA285696.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202623 | AK047778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202623 | BC046619.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202623 | NM_028832.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202623 | AA285696.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202623 | AK047778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202623 | BC046619.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27306691 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145706 | - | - |
| Whole Mouse Genome G4122A | A_51_P202623 | NM_028832.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202623 | AA285696.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202623 | AK047778.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202623 | BC046619.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202623 | NM_028832.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202623 | AA285696.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202623 | AK047778.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202623 | BC046619.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139198 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145706 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08353147 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08353166 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08353176 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08353170 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08353168 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08353175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27306673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08874369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08781349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08781344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08351102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08351100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08947614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08881652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08881655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08353149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08966511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08966539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08853469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08881654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08966531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08781345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27306648 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10371397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371399 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371398 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455700_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10365420 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10371396 | - | - |
| Mouse Genome 430 2.0 Array | 1455700_at | - | - |
| Mouse Genome 430 2.0 Array | 1455700_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74238_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74238_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000019614.mm.1 | NM_028832.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000019614.mm.1 | NM_028832.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000019614.mm.1 | TC1000002604.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455700_at | - | - |
| Mouse Expression Set 430 | 1455700_PM_at | - | - |
| Murine Genome U74v2 Set | 97107_PM_at | - | - |
| Murine Genome U74v2 Set | 97107_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455700_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455700_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455700_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455700_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 491539 | - | - |
| ABI Mouse Genome Survey Microarray | 756264 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4810594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4810594 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810594 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2605803 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213317 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2605803 | - | - |
| mouse-6 v1.1 (Illumina) | scl37671.3.1_35-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810594 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37671.3.1_35-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810594 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2997382 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2997379 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2997379 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2997382 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37671.3.1_35-S | - | - |

Mterf2
Genomic Map