Assay Details
Gene Symbol: Ccny | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026484.3 | NP_080760.2 | 4 - 5 | 848 | 79 | |
GenBank mRNA | AK017493.1 | - | 4 - 5 | 680 | 79 | |
AK028696.1 | - | 4 - 5 | 849 | 79 | ||
AK076556.1 | - | 4 - 5 | 378 | 79 | ||
AK215714.1 | - | 2 - 3 | 142 | 79 | ||
BC023321.1 | - | 5 - 6 | 766 | 3995040 | 79 | |
BC035524.1 | - | 3 - 4 | 603 | 4974858 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
