Assay Details
| Gene Symbol: Ccny | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026484.3 | NP_080760.2 | 2 - 3 | 704 | 82 | |
| GenBank mRNA | AK017493.1 | - | 2 - 3 | 536 | 82 | |
| AK028696.1 | - | 2 - 3 | 705 | 82 | ||
| AK076556.1 | - | 2 - 3 | 234 | 82 | ||
| BC023321.1 | - | 3 - 4 | 622 | 3995040 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccny
Genomic Map