Assay Details
| Gene Symbol: Rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK152122.1 | - | 4 - 5 | 806 | 71 | |
| S79539.1 | - | 2 - 3 | 132 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the S79539.1 transcripts.
Set Membership: |
|

Rab3ip
Genomic Map