Assay Details
| Gene Symbol: Rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003950.2 | NP_001003950.1 | 5 - 6 | 947 | 77 | |
| GenBank mRNA | AK152122.1 | - | 6 - 7 | 1094 | 77 | |
| AK206753.1 | - | 1 - 2 | 167 | 77 | ||
| BC080289.1 | - | 5 - 6 | 947 | 6816077 | 77 | |
| S79539.1 | - | 3 - 4 | 1027 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab3ip
Genomic Map