Assay Details
| Gene Symbol: Rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003950.2 | NP_001003950.1 | 6 - 7 | 1070 | 78 | |
| GenBank mRNA | AK152122.1 | - | 7 - 8 | 1217 | 78 | |
| AK209726.1 | - | 1 - 2 | 66 | 78 | ||
| BC080289.1 | - | 6 - 7 | 1070 | 6816077 | 78 | |
| S79539.1 | - | 4 - 5 | 1150 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab3ip
Genomic Map