Assay Details
| Gene Symbol: Rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003950.2 | NP_001003950.1 | 7 - 8 | 1183 | 78 | |
| GenBank mRNA | AK152122.1 | - | 8 - 9 | 1330 | 78 | |
| BC080289.1 | - | 7 - 8 | 1183 | 6816077 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab3ip
Genomic Map