Assay Details
| Gene Symbol: Rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003950.2 | NP_001003950.1 | 8 - 9 | 1282 | 90 | |
| GenBank mRNA | AK152122.1 | - | 9 - 10 | 1429 | 90 | |
| BC080289.1 | - | 8 - 9 | 1282 | 6816077 | 90 | |
| S79539.1 | - | 6 - 7 | 1359 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab3ip
Genomic Map