Assay Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025602.3 | NP_001020773.1 | 6 - 7 | 1048 | 75 | |
| NM_001294171.1 | NP_001281100.1 | 6 - 7 | 881 | 75 | ||
| NM_010743.3 | NP_034873.2 | 6 - 7 | 1048 | 75 | ||
| GenBank mRNA | AK033417.1 | - | 6 - 7 | 901 | 75 | |
| AK145118.1 | - | 6 - 7 | 902 | 75 | ||
| BC051445.1 | - | 6 - 7 | 858 | 30073421 | 75 | |
| D13695.1 | - | 5 - 6 | 740 | 75 | ||
| M24843.1 | - | 6 - 7 | 889 | 75 | ||
| Y07519.1 | - | 6 - 7 | 892 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il1rl1
Genomic Map