Assay Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025602.3 | NP_001020773.1 | 8 - 9 | 1340 | 135 | |
| GenBank mRNA | AK145118.1 | - | 8 - 9 | 1194 | 135 | |
| D13695.1 | - | 7 - 8 | 1032 | 135 | ||
More Information
Set Membership: |
|

Il1rl1
Genomic Map