Assay Details
| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018383.1 | - | 1 - 2 | 164 | 87 | |
| AK079735.1 | - | 171 | 87 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem19| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018383.1 | - | 1 - 2 | 164 | 87 | |
| AK079735.1 | - | 171 | 87 | |||
Set Membership: |
|