Assay Details
| Gene Symbol: Ankrd23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153502.3 | NP_705722.2 | 8 - 9 | 851 | 69 | |
| GenBank mRNA | AF492400.1 | - | 8 - 9 | 851 | 69 | |
| BC023221.1 | - | 8 - 9 | 828 | 69 | ||
| BC132370.1 | - | 8 - 9 | 861 | 40130647 | 69 | |
| BC138797.1 | - | 8 - 9 | 861 | 8861819 | 69 | |
| BC145421.1 | - | 7 - 8 | 762 | 9053979 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052882 | BC138797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052882 | BC145421.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052882 | BC023221.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052882 | BC132370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052882 | NM_153502.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047986 | BC022973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047986 | BC023221.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047986 | NM_153502.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047986 | AF492400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047986 | AK004293.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666041 | AK132827.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666041 | NM_001033799.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269723 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840669 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 81814 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052882 | BC138797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052882 | BC145421.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052882 | BC023221.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052882 | BC132370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052882 | NM_153502.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047986 | BC022973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047986 | BC023221.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047986 | NM_153502.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047986 | AF492400.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047986 | AK004293.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393223 | BC022973.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393223 | BC023221.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393223 | AK004293.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20148895 | - | - |
| Whole Mouse Genome G4122A | A_51_P393223 | BC022973.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393223 | BC023221.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393223 | AK004293.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P380887 | BC022973.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P380887 | BC023221.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P380887 | NM_153502.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P380887 | AF492400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P380887 | AK004293.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122146 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00635190 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00635191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00635198 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00635200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20148847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00635200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00635194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00496849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00494899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00618604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00635197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00796109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01066913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00618601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00635190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00635198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00618605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20148852 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10353888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353879 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353880 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353882 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353884 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353890 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353883 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353881 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353885 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428183_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10353878 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10353891 | - | - |
| Mouse Genome 430 2.0 Array | 1428183_at | - | - |
| Mouse Genome 430 2.0 Array | 1428183_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78321_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78321_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100017490.mm.1 | NM_153502.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017490.mm.1 | TC0100003874.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100017488.mm.1 | NM_153502.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100017488.mm.1 | TC0100003874.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428183_PM_at | - | - |
| Mouse Expression Set 430 | 1428183_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428183_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428183_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 736636 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1780164 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1780164 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001780164 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2741117 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2741117 | - | - |
| mouse-6 v1.1 (Illumina) | scl078321.1_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1780164 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl078321.1_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1780164 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2741117 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2741117 | - | - |

Ankrd23
Genomic Map