Assay Details
| Gene Symbol: Fshr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013523.3 | NP_038551.3 | 8 - 9 | 730 | 127 | |
| GenBank mRNA | AF095642.1 | - | 8 - 9 | 667 | 127 | |
| AK016635.1 | - | 8 - 9 | 730 | 127 | ||
| BC137990.1 | - | 8 - 9 | 727 | 8861009 | 127 | |
| BC137991.1 | - | 8 - 9 | 727 | 8861010 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fshr
Genomic Map