Assay Details
| Gene Symbol: Cpne4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028719.1 | NP_082995.1 | 8 - 9 | 1092 | 103 | |
| GenBank mRNA | AK043692.1 | - | 8 - 9 | 1105 | 103 | |
| AK191212.1 | - | 1 - 2 | 108 | 103 | ||
| BC043087.1 | - | 8 - 9 | 1092 | 6417611 | 103 | |
| BC063081.1 | - | 8 - 9 | 1092 | 6417611 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpne4
Genomic Map