Assay Details
| Gene Symbol: Cpne4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028719.1 | NP_082995.1 | 10 - 11 | 1239 | 93 | |
| GenBank mRNA | AK043692.1 | - | 10 - 11 | 1252 | 93 | |
| BC043087.1 | - | 10 - 11 | 1239 | 6417611 | 93 | |
| BC063081.1 | - | 10 - 11 | 1239 | 6417611 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpne4
Genomic Map