Assay Details
| Gene Symbol: Cpne4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028719.1 | NP_082995.1 | 3 - 4 | 670 | 72 | |
| GenBank mRNA | AK014396.1 | - | 3 - 4 | 672 | 72 | |
| AK043692.1 | - | 3 - 4 | 683 | 72 | ||
| AK202965.1 | - | 1 - 2 | 36 | 72 | ||
| BC043087.1 | - | 3 - 4 | 670 | 6417611 | 72 | |
| BC063081.1 | - | 3 - 4 | 670 | 6417611 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpne4
Genomic Map