Assay Details
| Gene Symbol: Cpne4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028719.1 | NP_082995.1 | 6 - 7 | 907 | 61 | |
| GenBank mRNA | AK014396.1 | - | 6 - 7 | 909 | 61 | |
| AK043692.1 | - | 6 - 7 | 920 | 61 | ||
| AK202965.1 | - | 4 - 5 | 273 | 61 | ||
| BC043087.1 | - | 6 - 7 | 907 | 6417611 | 61 | |
| BC063081.1 | - | 6 - 7 | 907 | 6417611 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpne4
Genomic Map