Assay Details
| Gene Symbol: Cldn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194921.1 | NP_001181850.1 | 4 - 5 | 674 | 57 | |
| NM_019815.3 | NP_062789.1 | 4 - 5 | 737 | 57 | ||
| GenBank mRNA | AA028634.1 | - | 2 - 3 | 189 | 57 | |
| AF221068.1 | - | 4 - 5 | 689 | 57 | ||
| AF349451.1 | - | 4 - 5 | 675 | 57 | ||
| AK033657.1 | - | 4 - 5 | 675 | 57 | ||
More Information
Set Membership: |
|

Cldn18
Genomic Map