Assay Details
| Gene Symbol: Iqcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028843.1 | NP_083119.1 | 2 - 3 | 179 | 101 | |
| GenBank mRNA | AK006797.1 | - | 2 - 3 | 155 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193296 | NM_028843.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193296 | AK006797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193296 | BC049769.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193296 | NM_001146701.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193296 | NM_028843.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193296 | AK006797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193296 | BC049769.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P193296 | NM_001146701.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671695 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106185 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143003 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193296 | NM_028843.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193296 | AK006797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193296 | BC049769.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193296 | NM_001146701.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193296 | NM_028843.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193296 | AK006797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193296 | BC049769.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P193296 | NM_001146701.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143003 | - | - |
| Whole Mouse Genome G4122A | A_51_P193296 | NM_028843.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193296 | AK006797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193296 | BC049769.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193296 | NM_001146701.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193296 | NM_028843.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193296 | AK006797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193296 | BC049769.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193296 | NM_001146701.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193291 | NM_028843.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193291 | AK006797.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138880 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07954633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26803755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07475203 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10588539 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588541 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430002_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10588538 | - | - |
| Mouse Genome 430 2.0 Array | 1430002_at | - | - |
| Mouse Genome 430 2.0 Array | 1430002_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74267_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74267_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900010571.mm.1 | NM_001146701.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010571.mm.1 | NM_001146701.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010571.mm.1 | NM_028843.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010571.mm.1 | NM_028843.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010571.mm.1 | TC0900001351.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430002_PM_at | - | - |
| Mouse Expression Set 430 | 1430002_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430002_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430002_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 825574 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100730411 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234948 | - | - |

Iqcf1
Genomic Map