Assay Details
| Gene Symbol: Tmem247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277980.1 | NP_001264909.1 | 1 - 2 | 235 | 63 | |
| NM_030104.1 | NP_084380.1 | 1 - 2 | 235 | 63 | ||
| GenBank mRNA | AK018916.1 | - | 1 - 2 | 169 | 63 | |
| AK076910.1 | - | 1 - 2 | 146 | 63 | ||
| BC100486.1 | - | 1 - 2 | 149 | 63 | ||
| CN837600.1 | - | 236 | 63 | |||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486719 | NM_030104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486719 | AK018916.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486719 | AK076910.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486719 | BC100486.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486719 | NM_030104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486719 | AK018916.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486719 | AK076910.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P486719 | BC100486.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145430 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894201 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938146 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128103 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139145 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486719 | NM_030104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486719 | AK018916.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486719 | AK076910.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486719 | BC100486.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486719 | NM_030104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486719 | AK018916.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486719 | AK076910.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P486719 | BC100486.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139145 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13096858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13499799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31419989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13191508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13280492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13313019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31419996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13199882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13096853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13499812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31419992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13392719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13249055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31419995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13145890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31419993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31419987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31420003 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10447340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447338 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447339 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439564_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10447337 | - | - |
| Mouse Genome 430 2.0 Array | 1439564_at | - | - |
| Mouse Genome 430 2.0 Array | 1439564_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78469_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78469_at | - | - |
| Mouse Expression Set 430 | 1439564_PM_at | - | - |
| Mouse Expression Set 430 | 1439564_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439564_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439564_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3610064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246883 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246883 | - | - |

Tmem247
Genomic Map