Assay Details
| Gene Symbol: Socs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033206.1 | - | 3 - 4 | 1062 | 82 | |
| AK048608.1 | - | 1 - 2 | 2437 | 82 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Socs2| Gene Symbol: Socs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033206.1 | - | 3 - 4 | 1062 | 82 | |
| AK048608.1 | - | 1 - 2 | 2437 | 82 | ||
Set Membership: |
|