Assay Details
| Gene Symbol: Slc22a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009203.3 | NP_033229.3 | 2 - 3 | 825 | 84 | |
| GenBank mRNA | AB005451.1 | - | 2 - 3 | 606 | 84 | |
| AK142580.1 | - | 2 - 3 | 143 | 84 | ||
| BC035927.1 | - | 761 | 4242354 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a12
Genomic Map