Assay Details
| Gene Symbol: Amdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027908.1 | NP_082184.1 | 3 - 4 | 374 | 62 | |
| GenBank mRNA | AK005066.1 | - | 3 - 4 | 330 | 62 | |
| AK090143.1 | - | 3 - 4 | 364 | 62 | ||
| BB856590.1 | - | 3 - 4 | 376 | 62 | ||
| BC022626.1 | - | 3 - 4 | 252 | 62 | ||
| BC043681.1 | - | 3 - 4 | 307 | 62 | ||
| BC055025.1 | - | 3 - 4 | 272 | 62 | ||
| BC118616.1 | - | 3 - 4 | 361 | 40107296 | 62 | |
| BC121826.1 | - | 3 - 4 | 360 | 40107295 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amdhd1
Genomic Map