Assay Details
| Gene Symbol: Amdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC022626.1 | - | 6 - 7 | 880 | 95 | |
| BC043681.1 | - | 6 - 7 | 935 | 95 | ||
| BC055025.1 | - | 6 - 7 | 900 | 95 | ||
| BC118616.1 | - | 6 - 7 | 989 | 40107296 | 95 | |
| BC121826.1 | - | 6 - 7 | 988 | 40107295 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Amdhd1
Genomic Map