Assay Details
| Gene Symbol: Spdya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142631.1 | NP_001136103.1 | 5 - 6 | 650 | 94 | |
| NM_029254.1 | NP_083530.1 | 5 - 6 | 650 | 94 | ||
| GenBank mRNA | AK014910.1 | - | 5 - 6 | 640 | 94 | |
| AK077138.1 | - | 5 - 6 | 651 | 94 | ||
| AY820306.1 | - | 4 - 5 | 551 | 94 | ||
| AY820307.1 | - | 4 - 5 | 551 | 94 | ||
| BC147706.1 | - | 5 - 6 | 628 | 9087465 | 94 | |
| BC147719.1 | - | 5 - 6 | 628 | 9087478 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P671722 | AK077138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P671722 | NM_001142631.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P671722 | AK077138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P671722 | NM_001142631.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P118225 | NM_029254.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P118225 | AK014910.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P118225 | CJ209011.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P118225 | AK004173.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P118225 | NR_027953.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391807 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130366 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917739 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 642445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263217 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53783 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924253 | AB030442.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924253 | AB043953.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924253 | AF168364.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924253 | AK042709.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924253 | AK085885.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924253 | AK155165.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924253 | AK172072.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924253 | BC113203.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924253 | NM_013781.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640230 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774510 | XM_484066.4 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774510 | XM_915478.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865462 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73230 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57386 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 143471 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 158207 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163757 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150969 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136354 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P671722 | AK077138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P671722 | NM_001142631.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P671722 | AK077138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P671722 | NM_001142631.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P118225 | NM_029254.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P118225 | AK014910.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P118225 | CJ209011.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P118225 | AK004173.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P118225 | NR_027953.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150969 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136354 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03363611 | - | - |
| Whole Mouse Genome G4122A | A_52_P671722 | AK077138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P671722 | NM_001142631.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P671722 | AK077138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P671722 | NM_001142631.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118225 | NM_029254.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118225 | AK014910.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118225 | CJ209011.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118225 | AK004173.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118225 | NR_027953.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136354 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13219706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13087800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31334993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13322468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13354605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13108399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31334998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13443086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31334995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13148596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13344620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13108393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13376849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13137822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31335004 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10446689 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446687 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446690 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446691 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446685 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446684 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446688 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429470_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441390_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10446679 | - | - |
| Mouse Genome 430 2.0 Array | 1437481_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432134_at | - | - |
| Mouse Genome 430 2.0 Array | 1432135_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1441390_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441390_at | - | - |
| Mouse Genome 430 2.0 Array | 1437481_at | - | - |
| Mouse Genome 430 2.0 Array | 1432134_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432135_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429470_at | - | - |
| Mouse Genome 430 2.0 Array | 1429470_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70891_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70891_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700010712.mm.1 | NM_001142631.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700010712.mm.1 | NM_001142631.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700010712.mm.1 | NM_029254.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700010712.mm.1 | NM_029254.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700010712.mm.1 | TC1700001137.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700010713.mm.1 | NM_001142631.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700010713.mm.1 | NM_001142631.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700010713.mm.1 | TC1700001137.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700010714.mm.1 | NM_001142631.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700010714.mm.1 | NM_001142631.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700010714.mm.1 | TC1700001137.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429470_PM_at | - | - |
| Mouse Expression Set 430 | 1441390_PM_at | - | - |
| Mouse Expression Set 430 | 1441390_at | - | - |
| Mouse Expression Set 430 | 1432135_s_at | - | - |
| Mouse Expression Set 430 | 1432135_PM_s_at | - | - |
| Mouse Expression Set 430 | 1429470_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429470_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441390_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429470_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441390_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 727750 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 102650095 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224739 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213006 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102650095 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102650095 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2545271 | - | - |

Spdya
Genomic Map