Assay Details
| Gene Symbol: Emilin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145158.3 | NP_660140.1 | 1 - 2 | 254 | 80 | |
| GenBank mRNA | AF468645.1 | - | 1 - 2 | 205 | 80 | |
| AK154230.1 | - | 1 - 2 | 224 | 80 | ||
| AK155037.1 | - | 1 - 2 | 232 | 80 | ||
| AK155306.1 | - | 1 - 2 | 248 | 80 | ||
| AK155366.1 | - | 1 - 2 | 235 | 80 | ||
| AK155554.1 | - | 1 - 2 | 250 | 80 | ||
| AK155797.1 | - | 1 - 2 | 251 | 80 | ||
| AK155878.1 | - | 1 - 2 | 252 | 80 | ||
| AK155879.1 | - | 1 - 2 | 248 | 80 | ||
| AK155909.1 | - | 1 - 2 | 254 | 80 | ||
| AK155913.1 | - | 1 - 2 | 224 | 80 | ||
| AK155918.1 | - | 1 - 2 | 281 | 80 | ||
| AK170081.1 | - | 1 - 2 | 251 | 80 | ||
| AK170288.1 | - | 1 - 2 | 224 | 80 | ||
| AK170391.1 | - | 1 - 2 | 224 | 80 | ||
| AK170801.1 | - | 1 - 2 | 255 | 80 | ||
| AK170822.1 | - | 1 - 2 | 276 | 80 | ||
| AK170953.1 | - | 1 - 2 | 270 | 80 | ||
| AK171015.1 | - | 1 - 2 | 276 | 80 | ||
| AK171020.1 | - | 1 - 2 | 242 | 80 | ||
| AK171309.1 | - | 1 - 2 | 251 | 80 | ||
| AK171579.1 | - | 1 - 2 | 224 | 80 | ||
| AK171592.1 | - | 1 - 2 | 224 | 80 | ||
| AK171616.1 | - | 1 - 2 | 224 | 80 | ||
| BC053753.1 | - | 1 - 2 | 538 | 6510220 | 80 | |
| GQ872457.1 | - | 1 - 2 | 149 | 80 | ||
| GQ872460.1 | - | 1 - 2 | 149 | 80 | ||
More Information
Set Membership: |
|

Emilin2
Genomic Map