| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700022850.mm.1 |
NM_145158.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022850.mm.1 |
NM_145158.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022850.mm.1 |
TC1700002490.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022849.mm.1 |
NM_145158.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022849.mm.1 |
NM_145158.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022849.mm.1 |
TC1700002490.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022848.mm.1 |
NM_145158.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022848.mm.1 |
NM_145158.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022848.mm.1 |
TC1700002490.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022847.mm.1 |
NM_145158.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022847.mm.1 |
NM_145158.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022847.mm.1 |
TC1700002490.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022851.mm.1 |
TC1700002490.mm.1 |
MTA TC |