Assay Details
| Gene Symbol: Cd248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054042.2 | NP_473383.1 | 1 | 2524 | 59 | |
| GenBank mRNA | AF378758.1 | - | 1 | 2482 | 59 | |
| AF388572.1 | - | 1 | 2479 | 59 | ||
| AK144655.1 | - | 1 | 2494 | 59 | ||
| BC046318.1 | - | 1 | 2524 | 6314798 | 59 | |
| BC128295.1 | - | 1 | 621 | 59 | ||
| BC152362.1 | - | 1 | 621 | 59 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926690 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926693 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926694 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926692 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188271 | AK144655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188271 | BC046318.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188271 | BC128295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188271 | BC152362.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188271 | AF378758.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188271 | AK144655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188271 | BC046318.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188271 | BC128295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188271 | BC152362.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P188271 | AF378758.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176274 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188271 | AK144655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188271 | BC046318.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188271 | BC128295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188271 | BC152362.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188271 | AF378758.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188271 | AK144655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188271 | BC046318.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188271 | BC128295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188271 | BC152362.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P188271 | AF378758.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P5454 | BC046318.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P5454 | BC128295.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P5454 | BC152362.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P5454 | AF378758.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P5454 | AF388572.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188271 | AK144655.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188271 | BC046318.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188271 | BC128295.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188271 | BC152362.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188271 | AF378758.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188271 | AK144655.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188271 | BC046318.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188271 | BC128295.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188271 | BC152362.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P188271 | AF378758.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926698 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176274 | - | - |
| Whole Mouse Genome G4122A | A_52_P5454 | BC046318.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5454 | BC128295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5454 | BC152362.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5454 | AF378758.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P5454 | AF388572.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188271 | AK144655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188271 | BC046318.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188271 | BC128295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188271 | BC152362.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188271 | AF378758.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188271 | AK144655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188271 | BC046318.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188271 | BC128295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188271 | BC152362.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188271 | AF378758.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106942 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119890 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176274 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14441152 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14441154 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14441156 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14441159 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14441161 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926692 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926694 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926690 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14336622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14490518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14365875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14338026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14338033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14490527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14245375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14490496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14490492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14398259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14490483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14365884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14338014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14338010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14317961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14441163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14338020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14258723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14365874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926740 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460543 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460542 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417439_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460541 | - | - |
| Mouse Genome 430 2.0 Array | 1417439_at | - | - |
| Mouse Genome 430 2.0 Array | 1417439_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70445_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70445_at | - | - |
| Mouse Expression Set 430 | 1417439_at | - | - |
| Mouse Expression Set 430 | 1417439_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417439_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417439_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417439_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417439_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2480605 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2480605 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002480605 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2670001 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2670001 | - | - |
| mouse-6 v1.1 (Illumina) | scl52830.2.1_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2480605 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52830.2.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2896843 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2670001 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2896843 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2670001 | - | - |

Cd248
Genomic Map