| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00017629.mm.1 |
NM_001077354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017629.mm.1 |
NM_001077354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017629.mm.1 |
TC0X00002794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017624.mm.1 |
NM_001077354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017624.mm.1 |
NM_001077354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017624.mm.1 |
TC0X00002794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017628.mm.1 |
NM_001077354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017628.mm.1 |
NM_001077354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017628.mm.1 |
TC0X00002794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017630.mm.1 |
NM_001077354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017630.mm.1 |
NM_001077354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017630.mm.1 |
TC0X00002794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017632.mm.1 |
NM_001077354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017632.mm.1 |
NM_001077354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017632.mm.1 |
TC0X00002794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017623.mm.1 |
NM_001077354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017623.mm.1 |
NM_001077354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017623.mm.1 |
TC0X00002794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017631.mm.1 |
NM_001077354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017631.mm.1 |
NM_001077354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017631.mm.1 |
TC0X00002794.mm.1 |
MTA TC |