Assay Details
| Gene Symbol: Cpsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018813.3 | NP_061283.2 | 5 - 6 | 718 | 83 | |
| GenBank mRNA | AF203969.1 | - | 5 - 6 | 571 | 83 | |
| AK149953.1 | - | 5 - 6 | 645 | 83 | ||
| BC100569.1 | - | 5 - 6 | 616 | 30041540 | 83 | |
| CF725936.1 | - | 3 - 4 | 327 | 83 | ||
More Information
Set Membership: |
|

Cpsf3
Genomic Map