Assay Details
| Gene Symbol: Cpsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018813.3 | NP_061283.2 | 6 - 7 | 809 | 90 | |
| GenBank mRNA | BC100569.1 | - | 6 - 7 | 707 | 30041540 | 90 |
| CF725936.1 | - | 4 - 5 | 418 | 90 | ||
More Information
Set Membership: |
|

Cpsf3
Genomic Map