Assay Details
| Gene Symbol: Kcnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008430.2 | NP_032456.2 | 1 - 2 | 709 | 99 | |
| GenBank mRNA | BC003729.1 | - | 1 - 2 | 406 | 3592756 | 99 |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnk1
Genomic Map