Assay Details
| Gene Symbol: Tubb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080971.2 | NP_001074440.1 | 1 - 2 | 208 | 72 | |
| GenBank mRNA | BC147322.1 | - | 1 - 2 | 106 | 9056380 | 72 |
| BC147323.1 | - | 1 - 2 | 106 | 9056384 | 72 | |
| BC147687.1 | - | 1 - 2 | 208 | 9056660 | 72 | |
| BC147698.1 | - | 1 - 2 | 208 | 9056671 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tubb1
Genomic Map