Assay Details
| Gene Symbol: Tubb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080971.2 | NP_001074440.1 | 3 - 4 | 427 | 64 | |
| GenBank mRNA | BC147322.1 | - | 3 - 4 | 325 | 9056380 | 64 |
| BC147323.1 | - | 3 - 4 | 325 | 9056384 | 64 | |
| BC147687.1 | - | 3 - 4 | 427 | 9056660 | 64 | |
| BC147698.1 | - | 3 - 4 | 427 | 9056671 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P126159 | BC147687.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P126159 | BC147698.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P126159 | NM_001080971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P126159 | BC147322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P126159 | BC147323.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499281 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34790 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P126159 | BC147687.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P126159 | BC147698.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P126159 | NM_001080971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P126159 | BC147322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P126159 | BC147323.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01439290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01567080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01547225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01413843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02127954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01413840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01248956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01567060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01167068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01439278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01567073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01439288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875485 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10479157 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10479155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10479158 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10479156 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444214_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10479154 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200022161.mm.1 | NM_001080971.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200022161.mm.1 | TC0200002706.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200022160.mm.1 | NM_001080971.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200022160.mm.1 | TC0200002706.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200022162.mm.1 | NM_001080971.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200022162.mm.1 | TC0200002706.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200022163.mm.1 | NM_001080971.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200022163.mm.1 | TC0200002706.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1444214_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444214_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2987747 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2987747 | - | - |

Tubb1
Genomic Map