| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200018361.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018361.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018361.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018364.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018364.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018364.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018365.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018365.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018365.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018370.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018370.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018370.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018362.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018362.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018362.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018368.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018368.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018368.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018367.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018367.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018367.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018358.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018358.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018358.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018359.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018359.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018359.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018360.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018360.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018360.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018366.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018366.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018366.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018363.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018363.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018363.mm.1 |
TC0200002328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018369.mm.1 |
NM_199303.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018369.mm.1 |
NM_199303.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018369.mm.1 |
TC0200002328.mm.1 |
MTA TC |