Assay Details
| Gene Symbol: Usp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206851.1 | NP_001193780.1 | 5 - 6 | 1971 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp44| Gene Symbol: Usp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206851.1 | NP_001193780.1 | 5 - 6 | 1971 | 84 | |
Set Membership: |
|