Assay Details
| Gene Symbol: Crhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007762.4 | NP_031788.1 | 1 - 2 | 222 | 73 | |
| GenBank mRNA | AK144430.1 | - | 1 - 2 | 291 | 73 | |
| AK158806.1 | - | 1 - 2 | 223 | 73 | ||
| BC103675.1 | - | 1 - 2 | 50 | 40046992 | 73 | |
| BC104735.1 | - | 1 - 2 | 50 | 40046995 | 73 | |
| BC105673.1 | - | 1 - 2 | 50 | 40046994 | 73 | |
| BC105675.1 | - | 1 - 2 | 50 | 40046993 | 73 | |
| X72305.1 | - | 1 - 2 | 63 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158806.1, BC103675.1, BC104735.1, BC105673.1, BC105675.1, NM_007762.4, X72305.1 transcripts.
Set Membership: |
|

Crhr1
Genomic Map