Assay Details
| Gene Symbol: Crhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007762.4 | NP_031788.1 | 2 - 3 | 307 | 73 | |
| GenBank mRNA | AB375515.1 | - | 2 - 3 | 117 | 73 | |
| AF483484.1 | - | 2 - 3 | 117 | 73 | ||
| AF483485.1 | - | 2 - 3 | 117 | 73 | ||
| AK144430.1 | - | 2 - 3 | 375 | 73 | ||
| AK158806.1 | - | 2 - 3 | 308 | 73 | ||
| BC103675.1 | - | 2 - 3 | 135 | 40046992 | 73 | |
| BC104735.1 | - | 2 - 3 | 135 | 40046995 | 73 | |
| BC105673.1 | - | 2 - 3 | 135 | 40046994 | 73 | |
| BC105675.1 | - | 2 - 3 | 135 | 40046993 | 73 | |
| X72305.1 | - | 2 - 3 | 148 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105673.1 transcripts.
Set Membership: |
|

Crhr1
Genomic Map