Assay Details
| Gene Symbol: Crhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007762.4 | NP_031788.1 | 5 - 6 | 627 | 67 | |
| GenBank mRNA | AB375515.1 | - | 5 - 6 | 437 | 67 | |
| AF483484.1 | - | 5 - 6 | 437 | 67 | ||
| AF483485.1 | - | 5 - 6 | 437 | 67 | ||
| AK144430.1 | - | 5 - 6 | 695 | 67 | ||
| AK158806.1 | - | 5 - 6 | 628 | 67 | ||
| BC103675.1 | - | 5 - 6 | 455 | 40046992 | 67 | |
| BC104735.1 | - | 5 - 7 | 455 | 40046995 | 67 | |
| BC105673.1 | - | 6 - 7 | 531 | 40046994 | 67 | |
| BC105675.1 | - | 5 - 6 | 455 | 40046993 | 67 | |
| FJ668671.1 | - | 4 - 5 | 317 | 67 | ||
| X72305.1 | - | 5 - 6 | 468 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105673.1 transcripts.
Set Membership: |
|

Crhr1
Genomic Map