Assay Details
| Gene Symbol: Epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007884.2 | NP_031910.1 | 6 - 7 | 816 | 137 | |
| GenBank mRNA | AK015223.1 | - | 6 - 7 | 816 | 137 | |
| AK137179.1 | - | 5 - 6 | 741 | 137 | ||
| AK157900.1 | - | 1067 | 137 | |||
| AK157901.1 | - | 5 - 6 | 1078 | 137 | ||
| BC012695.1 | - | 5 - 6 | 788 | 4037028 | 137 | |
| D78274.1 | - | 5 - 6 | 731 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epyc
Genomic Map